Learn More
Labconco™ SpillStopper™ Work Surfaces for 4ft. Protector™XStream
Product Code. 15963117 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
No Cutouts
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15963117 No Cutouts Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15963117 Supplier Labconco™ Supplier No. 9503400
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
SpillStopper work surfaces are dished to contain spills
  • Front edge has large radius to aerodynamically direct airflow in the hood
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 3 x 6 in. cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately
Specifications
For Use With (Equipment) Protector XStream Fume Hood (48 in.)
Includes No Cutouts
Product Type Work Surface
Material Epoxy Resin
For Use With (Application) Laboratory Fume Hood Work