Learn More
Labconco™ XVS™ PVC Elbow for Ventilation Stations
Product Code. 15459444 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector VS Ventilation Station
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15459444 Protector VS Ventilation Station Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15459444 Supplier Labconco™ Supplier No. 4708700
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
PVC Elbow for Ventilation Stations
  • For use with Labconco Protector XVS Ventilation Stations
  • 90° elbow has belled end connections to receive PVC duct directly
Specifications
For Use With (Equipment) Protector VS Ventilation Station
Height (English) 13.62 in.
Weight (English) 8 lb.
Weight (Metric) 3.6 Kg
Height (Metric) 34.59 cm
Includes 90° Elbow
Dimensions 6 in. dia. Round PVC Duct
Material PVC
For Use With (Application) Designed for quick installation and minimum pressure losses