Learn More
Labconco™ Protector Glove Box Accessory Kits
Kits provided added functionality options for Protector glove boxes
10055.00 NOK
Specifications
For Use With (Equipment) Protector Glove Boxes
View More Specs
Products 1
Product Code Brand Includes Product Type Price Quantity & Availability  
Product Code Brand Includes Product Type Price Quantity & Availability  
16649183
View Documents Promotion Details
Labconco™
5441403
HEPA filter, hose barb connectors and 10 ft. of 0.75 in. ID neoprene tubing for attachment to the pressure relief bubbler exhaust Pressure Relief Kit
10055.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-11-2024
Description
Add-A-Valve Kits include two manual valves, tubes, nuts and instructions for adding valves to the glove box for evacuating with vacuum, filling with inert gas, connecting to a drying train, or connecting to filtration components. Manual valves mount in predrilled holes behind the label located below the control panel. For use with Protector Combination and Protector Filtered Glove Boxes.
Specifications
Specifications
Protector Glove Boxes