Learn More
Labconco™ Glass Trap Insert for CentriVap™ System
Product Code. 10492960
Click to view available options
Capacity (Metric):
2550 mL
Unit Size:
Each
Product Code. 10492960
Brand: Labconco™ 7397605
Only null left
Add to basket
Add to basket
Protects CentriVap Cold Trap reservoir and lid from corrosives.
  • Included with CentriVap Acid Systems; accessory for other systems
  • Not compatible with pre-2005 Cold Traps
Specifications
Type Trap Insert
Capacity (Metric) 2550 mL
No. of Shelves 0
Material Glass
For Use With (Equipment) CentriVap™ concentrator cold trap
Weight (English) 4 lb.
Weight (Metric) 1.8 kg