Learn More
Labconco™ FreeZone™ Six-Port Tray Dryer Manifold
Product Code. 11791449
Click to view available options
For Use With (Equipment):
Freezone™ Stoppering Tray Dryer
Unit Size:
Each
Product Code. 11791449
Brand: Labconco™ 7726500
Only null left
Add to basket
Add to basket
Six valves provide the flexibility to connect flasks and other freeze dry glassware to FreeZone Freeze Dry Systems with Tray Dryer.
  • Valves accommodate either 1/2 in. (1.3cm) or 3/4 in. (1.9cm) flask adapters
  • Mounts to support stand included with the Tray Dryer
  • May be field installed before or after the Tray Dryer is installed on the Freeze Dry System
Specifications
Type Tray Dryer Manifold
No. of Shelves 0
Material Stainless Steel
No. of Ports 6
For Use With (Equipment) Freezone™ Stoppering Tray Dryer
Width (English) 30 in.
Height (English) 7 in.
Height (Metric) 17.8 cm
Length (English) 16.5 in.
Length (Metric) 41.9 cm
Voltage 115 V
Electrical Requirements 115 V 50/60 Hz
Frequency 50/60 Hz
Weight (English) 225 lb.
Weight (Metric) 102 kg