Learn More
Labconco™ FreeZone™ Bulk Tray
Product Code. 11751449 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Freezone™ Bulk or Stoppering Tray Dryer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11751449 Freezone™ Bulk or Stoppering Tray Dryer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11751449 Supplier Labconco™ Supplier No. 7756200
  • ONLINE EXCLUSIVE PRICE
    4570.00 NOK
    4225.79 NOK / Pack of 1
    Save 344.21 NOK
    7% Off
 Restricted Item
Estimated Shipment: 17-02-2027
For FreeZone Bulk Tray Dryers and/or Stoppering Tray Dryers
  • Can be used with both Stoppering and Bulk Tray Dryers
Specifications
Type Bulk Tray
No. of Shelves 0
Material Stainless Steel
For Use With (Equipment) Freezone™ Bulk or Stoppering Tray Dryer
Width (English) 11.5 in.
Width (Metric) 29.2 cm
Height (English) 0.88 in.
Height (Metric) 2.2 cm
Length (English) 15 in.
Length (Metric) 38.1 cm
Weight (English) 3 lb.
Weight (Metric) 1.4 kg