Learn More
Labconco™ Exhaust Damper
Product Code. 15409434
Change view
Click to view available options
Diameter (English):
6 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) unitSize
15409434 6 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15409434 Supplier Labconco™ Supplier No. 3924000
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
For 2 ft., 3 ft., and 4 ft. enclosures.
Manual Duct Damper, powder-coated steel, 6 in. nominal diameter. For use with Labconco ventilated enclosures.
Specifications
For Use With (Equipment) Hoods and Enclosures
Weight (English) 1 lb.
Weight (Metric) 0.5 Kg
Product Type Exhaust Damper
Diameter (English) 6 in.
Material Powder-coated Steel
Diameter (Metric) 15.24 cm
For Use With (Application) Exhausting Enclosures