Learn More
Labconco™ Ducting Parts: Connectors
Product Code. 15789688 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Connector, Exhaust (Six Thimble)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15789688 Connector, Exhaust (Six Thimble) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15789688 Supplier Labconco™ Supplier No. 5440900
  • ONLINE EXCLUSIVE PRICE
    8565.00 NOK
    8156.03 NOK / Pack of 1
    Save 408.97 NOK
    4% Off
 Restricted Item
Estimated Shipment: 03-05-2027
Connectors for Labconco ducts
Powder-coated steel assembly for connecting a glove box to a building's exhaust or a remote blower. 6 in. diameter connection.
Specifications
For Use With (Equipment) Hoods and Enclosures
Product Type Connector, Exhaust (Six Thimble)