Learn More
Labconco™ DNA Rotor for CentriVap™ System
Description
Acid-Resistant Thermoplastic Rotors hold tubes at an angle to maximize surface area.
Acid-Resistant Thermoplastic Rotors hold tubes at an angle to maximize surface area. They are easily interchangeable. Sample containers are not included. For use with Labconco CentriVap Vacuum Concentrators.
Specifications
Specifications
| Material | Thermoplastic |
| Type | DNA Rotor |
| For Use With (Equipment) | CentriVap™ system |
| Holds | 56 Autosampler Vials |
| Shipping Weight (English) | 3 lb. |
| Shipping Weight (Metric) | 1.4 kg |
| Weight (English) | 3 lb. |
| Weight (Metric) | 1.4 kg |