Learn More
Labconco™ CentriVap™ -105 Cold Traps
Product Code. 12695345 Shop All Labconco Corporation Products
Change view
Click to view available options
Electrical Requirements:
230 V 50 Hz
Plug Type:
Schuko
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Plug Type Unit Size
12695345 230 V 50 Hz Schuko Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12695345 Supplier Labconco™ Supplier No. 7385030
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump from very low freezing point liquids
  • Brushed, stainless steel and glacier white, epoxy-coated steel exterior with blue accents;
  • Temperature graph illuminates green when collector coil reaches operating temperature of -65°C
  • HCFC-free refrigeration system with two 1/3 hp compressors lowers cold trap to -105°C (-157°F) in less than 40 min.
  • 4L stainless steel collector
  • Arylic lid with two Quick-Disconnect fittings for connection to a CentriVap Concentrator
  • Stainless steel drain port with corrosion-resistant, nylon valve
  • Grounding clip and ON/OFF power switch
Specifications
Certifications/Compliance CE
Material Stainless Steel
Type Benchtop Cold Trap
For Use With (Equipment) Very low freezing point solvent
Plug Type Schuko
Temperature (Metric) Range -105°C
Width (English) 24.1 in.
Width (Metric) 61.2 cm
Height (English) 17.6 in.
Height (Metric) 44.7 cm
Depth (English) 23.9 in.
Depth (Metric) 60.7 cm
Weight (English) 170 lb.
Weight (Metric) 77 kg
Electrical Requirements 230 V 50 Hz
Voltage 230 V
Frequency 50 Hz