Learn More
Labconco™ Automatic Pressure Controller Gas and Vacuum Tubing Connection Kit
Click to view available options
For Use With (Equipment):
Precise Basic Glove Box
Unit Size:
Each
Description
Include the flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box.
Specifications
Specifications
| For Use With (Equipment) | Precise Basic Glove Box |
| Weight (English) | 20 lb. |
| Weight (Metric) | 9.1 Kg |
| Product Type | Enclosure Kit |