missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Ammonia Trap Insert
Protects rotary vane vacuum pump
Brand: Labconco™ 7995600
Additional Details : Weight : 1.82000kg
Requires : Clear polypropylene canister with mounting bracket or clear polypropylene canister with stand for proper use
Description
Protects rotary vane vacuum pump
- For use with RapidVap™ Vacuum Evaporation System, Centrivap™ Concentrators or Glove Boxes
- Contains impregnated carbon
- Suitable for ammonia and low molecular weight amines
Specifications
Trap Insert | |
CentriVap™ concentrators, precise glove boxes, RapidVap™ vacuum evaporation system | |
4 lb. | |
1.8 kg |