Learn More
Labconco™ Labconco Activated Carbon Solvent Trap Insert for Pre-2017 Trap
Product Code. 15113095 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Activated Carbon Solvent Trap Insert
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15113095 Activated Carbon Solvent Trap Insert Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15113095 Supplier Labconco™ Supplier No. 7772600
  • ONLINE EXCLUSIVE PRICE
    6590.00 NOK
    5822.31 NOK / Pack of 1
    Save 767.69 NOK
    11% Off
 Restricted Item
Estimated Shipment: 15-02-2027
For use with 7772500 Activated Carbon Solvent Trap.
Traps are connected between the evaporator and vacuum pump to provide additional protection to the pump.
Specifications
Product Type Activated Carbon Solvent Trap Insert
Weight (English) 4.0 lb.
Weight (Metric) 1.8 kg