Learn More
Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X)
Product Code. 12673967 Shop All Epredia Products
Click to view available options
Quantity:
125 mL
1000 mL
Unit Size:
1000mL
125mL
Product Code. 12673967
Brand: Epredia™ TA125PT
Currently Unavailable: We have no information when this product will be available.
View alternative products
Employ a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X).
  • For in vitro diagnostic use
  • Added to promote more effective washings, resulting in decreased nonspecific background staining
  • Aids reagent spreading on slides
Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material PBS and Tween
Concentration 20X
For Use With (Application) Anatomical Pathology
pH 7.6
Quantity 125 mL
For in vitro diagnostic use