Learn More
SymLine Lab Sink with Drain
Product Code. 17937270
Click to view available options
Includes:
G 1.5 in. OD Thread
GL 25 Thread
Pipe 32 mm (OD), Angled
Pipe 32 mm (OD), Vertical
Unit Size:
Each
Product Code. 17937270
Brand: SymLine 118021
Only null left
Add to basket
Add to basket
Lab Sink with Drain for integration in fume cupboards or on worktops for easy disposal of liquid solvent waste in large quantities.
  • Made of dissipative PE-HD-EL
  • Base with integrated slope for complete emptying
  • Drain with removable sieve
Specifications
Color Black
Material HDPE
Type Laboratory Sink
Includes GL 25 Thread
Dimensions (L x W x H) 400 x 300 x 190 mm