Learn More
Elma™ Lab Clean N10 Ultrasonic Cleaning Solution
Safely cleans sensitive materials in an ultrasonic bath or automatic washer. Tovatech Elma Lab Clean N10 Cleaning Solution removes emulsions, marking and label residues, light grease and oil, fingerprintes and dust.
3790.00 NOK
Specifications
Product Type | Ultrasonic Cleaning Solution |
---|---|
Form | Liquid |
pH | Neutral |
Content And Storage | Keep between 43°F to 85°F, shelf life 2 years |
For Use With (Application) | Cleaning Any Sensitive Metal, Plastic, Glass or Ceramic Surface |
Alert:
Irritating to eyes.
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
13219662
|
Elma™
8000078 |
25 L | 1 Ea. |
3790.00 NOK
25 L |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Neutral pH general-purpose formulation
- Ideal for gentle cleaning of aluminum and light metals as well as any other material
- Residue-free - Leaves no residue after rinsing with DI water
- Dilute with water to 2% for use in an ultrasonic cleaner or to 1% for a labware washer
- Dilute with either tap water or deionized water
- Biodegradable
Applications
- For cleaning any sensitive metal, plastic, glass, or ceramic surface
- Cleans laboratory sieves, aluminum surfaces prior to anodizing or coating and laser machined parts
Specifications
Ultrasonic Cleaning Solution | |
Neutral | |
Cleaning Any Sensitive Metal, Plastic, Glass or Ceramic Surface |
Liquid | |
Keep between 43°F to 85°F, shelf life 2 years |