Learn More
L-Tryptophan methyl ester hydrochloride, 98%
Product Code. p-3212827 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
25g
5g
Product Code. 10493063
Brand: Thermo Scientific Acros 376650050
Request Bulk or Custom Format
CAS: 7524-52-9 | C12H15ClN2O2 | 254.71 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7524-52-9
Molecular Formula C12H15ClN2O2
Molecular Weight (g/mol) 254.71
MDL Number MFCD00066134
InChI Key XNFNGGQRDXFYMM-PPHPATTJSA-N
Synonym l-tryptophan methyl ester hydrochloride, h-trp-ome.hcl, methyl l-tryptophanate hydrochloride, methyl tryptophanate hydrochloride, methyl l-tryptophanate hcl, unii-au62o6861l, methyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, s-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, l-tryptophanmethylesterhydrochloride, l-tryptophan methyl ester hcl
PubChem CID 2734891
IUPAC Name methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC1=CNC2=CC=CC=C12
Specifications
Melting Point 218.0°C to 220.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic Bottle
Specific Rotation + 18.00
Specific Rotation Condition + 18.00 (20.00°C c=5 CH3OH)
Quantity 5 g
Formula Weight 254.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Tryptophan methyl ester hydrochloride