Learn More
Thermo Scientific Chemicals L-Tryptophan, Cell Culture Reagent
Product Code. p-7042434
Click to view available options
Quantity:
10 g
100 g
1 kg
Unit Size:
100g
10g
1kg
Product Code. 15414889
Brand: Thermo Scientific Chemicals J62508.09
Request Bulk or Custom Format
CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
Specifications
Melting Point 280°C to 285°C
Color White
Beilstein 86197
Merck Index 14,9797
Quantity 10 g
Formula Weight 204.23
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Tryptophan