missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine Allo-free base, 990%, MP Biomedicals™

Product Code. 11463451 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
This item is not returnable. View return policy

Product Code. 11463451

Brand: MP Biomedicals 0210305325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-threonine is classified as an essential amino acid. Synthesized from homoserine phosphate by threonine synthase or from acetaldehyde and glycine by serine hydroxymethyltransferase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
IUPAC Name 2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Color White
Assay Percent Range 99%
Quantity 25 g
Formula Weight 119.1
Percent Purity 99.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Threonine, Allo Free
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.