Learn More
Microspec™ L Shaped Spreaders
Product Code. 15732125
Change view
Click to view available options
Quantity:
500
7500
Product Type:
Inoculation Product
Spreader
Unit Size:
Pack of 500
Pack of 7500
Product Code. Quantity Product Type unitSize
15732125 7500 Spreader Pack of 7500
15742125 500 Inoculation Product Pack of 500
15752125 500 Spreader Pack of 500
3 options
Product Code. 15732125
Brand: Microspec™ E1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Features smooth edges which avoids tearing and digging into agar
  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Ideal for bacterial counts
Specifications
Disposable Single-use
Color Blue
Length (English) 5.5 in.
Length (Metric) 140 mm
Packaging Microseal Pouch, Ecopack
Quantity 7500
Material High-impact Polystyrene
Product Type Spreader
Sterility Aseptically Manufactured