missing translation for 'onlineSavingsMsg'
Learn More

L-Serinyl diphenylborinate, 95%, Thermo Scientific™

Catalog Number 15471688
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
16657-63-9
C15H16BNO3
269.11
MFCD22125144
KEEDBWSHNMLMJV-UHFFFAOYNA-N
diphenylboranyl 2s-2-amino-3-hydroxypropanoate, l-serinyl diphenylborinate
99738136
NC(CO)C(=O)OB(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
16657-63-9
C15H16BNO3
269.11
MFCD22125144
KEEDBWSHNMLMJV-UHFFFAOYNA-N
diphenylboranyl 2s-2-amino-3-hydroxypropanoate, l-serinyl diphenylborinate
99738136
NC(CO)C(=O)OB(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
IUPAC Name diphenylboranyl (2S)-2-amino-3-hydroxypropanoate
Formula Weight 269.11
Percent Purity 95%
Chemical Name or Material L-Serinyl diphenylborinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'