Learn More
Masterflex™ L/S™ Bulk-Packed Platinum-Cured Silicone High-Performance Precision Pump Tubing
Product Code. 11705788
Change view
Click to view available options
Tubing Size:
13
14
15
16
17
24
25
35
36
Flow Rate:
1000 mL/min
1100mL/min.
130 mL/min
1700 mL/min
1800 mL/min
2600 mL/min
3400 mL/min
36 mL/min
480 mL/min
Unit Size:
Pack of 10
Pack of 20
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Flow Rate unitSize
11705788 16 480 mL/min Pack of 20
11785778 13 36 mL/min Pack of 20
11795778 14 130 mL/min Pack of 20
11720359 17 1700 mL/min Pack of 20
11750359 24 1800 mL/min Pack of 20
11715788 25 1000 mL/min Pack of 20
11760359 35 2600 mL/min Pack of 10
11770359 36 3400 mL/min Pack of 10
11740359 15 1100mL/min. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11705788 Supplier Masterflex™ Supplier No. 9640416
Only null left
Add to basket
Add to basket
Excellent biocompatibility that meets multiple classifications
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
  • Pharmaceutical, biotech, peristaltic pumping, and food applications
  • Excellent heat resistance, low compression set, odorless, nontoxic, translucent
  • Comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size).
  • Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping.
  • Tubing comes in 25-ft bags.
Specifications
Material Platinum-cured Silicone
Overall Length (English) 25 ft.
Overall Length (Metric) 7.6 m
For Use With (Application) Pharmaceutical, Biotech, Peristaltic Pumping, and Food Applications
Min. Operating Temperature (Metric) -40°C
Tubing Size 16
Flow Rate 480 mL/min
Hose Barb 1/8 in.
Max. Operating Temperature (Metric) 100°C