Learn More
L-Praziquanamine, TRC
Specifications
Specifications
| Chemical Name or Material | L-Praziquanamine |
| CAS | 99746-73-3 |
| Molecular Formula | C12H14N2O |
| Quantity | 50 mg |
| Synonym | (11bS)-1,2,3,6,7,11b-Hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, (+)-Praziquanamine, (S)-1,2,3,6,7,11b-Hexahydro-4H-Pyrazino[2,1-a]isoquinolin-4-one, (11bS)-1,2,3,6,7,11b-Hexahydro-4H-Pyrazino[2,1-a]isoquinolin-4-one |
| InChI Formula | InChI=1S/C12H14N2O/c15-12-8-13-7-11-10-4-2-1-3-9(10)5-6-14(11)12/h1-4,11,13H,5-8H2/t11-/m1/s1 |
| SMILES | O=C1CNC[C@H]2N1CCc3ccccc23 |
| IUPAC Name | (11bS)-1,2,3,6,7,11b-hexahydropyrazino[2,1-a]isoquinolin-4-one |
| Molecular Weight (g/mol) | 202.26 |
| Formula Weight | 202.1106 |