Learn More
l-Methionine, Millipore™
Product Code. 10403791
Change view
Click to view available options
Quantity:
1 kg
10 kg
Unit Size:
10kg
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10403791 1 kg 1kg
10240701 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10403791 Supplier Millipore Supplier No. 1057071000
 Restricted Item
Estimated Shipment: 12-11-2026
For biochemistry
for biochemistry
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63-68-3
Specifications
Melting Point 280°C to 282°C
MDL Number MFCD00063097
Synonym L-Methionine, Met, L-Methionine
Density 1.34 g/cm3
Molecular Weight (g/mol) 149.2
Quantity 1 kg
Grade Research
Chemical Name or Material L-Methionine