Learn More
L-m-Tyrosine, TRC
Product Code. 30453534
Change view
Click to view available options
Quantity:
50 mg
250 mg
5 g
Unit Size:
250mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453534 250 mg 250mg
30451657 50 mg 50mg
30451655 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30453534 Supplier Toronto Research Chemicals Supplier No. TRCT910010250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 587-33-7
Molecular Formula C9 H11 N O3
Molecular Weight (g/mol) 181.19
Synonym L-Phenylalanine, 3-hydroxy-, m-Tyrosine, L- (8CI), 3-Hydroxy-L-phenylalanine, (2S)-2-Amino-3-(3-hydroxyphenyl)propanoic acid, (S)-2-Amino-3-(3-hydroxyphenyl)propanoic acid, 3-(m-Hydroxyphenyl)alanine, 3-Hydroxyphenylalanine, L-3-Hydroxyphenylalanine, L-m-Tyrosine, L-meta-Tyrosine, m-L-Tyrosine, (S)-α-Amino-β-(m-hydroxyphenyl)propionic acid
IUPAC Name (2S)-2-amino-3-(3-hydroxyphenyl)propanoic acid
SMILES N[C@@H](Cc1cccc(O)c1)C(=O)O
Specifications
InChI Formula InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-2-1-3-7(11)4-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1
Quantity 250 mg
Formula Weight 181.0739
Shipping Condition Room Temperature
Chemical Name or Material L-m-Tyrosine