Learn More
L-Lyxose, 99%, Thermo Scientific Chemicals
Catalog Number p-7025869
Click to view available options
Quantity:
1 g
5 g
1949-78-6
C5H10O5
150.13
MFCD00064111
PYMYPHUHKUWMLA-YUPRTTJUSA-N
l-lyxose, aldehydo-l-lyxose, lyxose, l, l-lyx, 2r,3r,4s-2,3,4,5-tetrahydroxypentanal, unii-m7j5hm9dy4, m7j5hm9dy4, l +-lyxose, l-lyxose 9ci
644176
CHEBI:28480
(2R,3R,4S)-2,3,4,5-tetrahydroxypentanal
C(C(C(C(C=O)O)O)O)O
1949-78-6
C5H10O5
150.13
MFCD00064111
PYMYPHUHKUWMLA-YUPRTTJUSA-N
l-lyxose, aldehydo-l-lyxose, lyxose, l, l-lyx, 2r,3r,4s-2,3,4,5-tetrahydroxypentanal, unii-m7j5hm9dy4, m7j5hm9dy4, l +-lyxose, l-lyxose 9ci
644176
CHEBI:28480
(2R,3R,4S)-2,3,4,5-tetrahydroxypentanal
C(C(C(C(C=O)O)O)O)O
It acts as a reducing carbohydrate present in maple syrup. It is used in molecular modeling calculations in the study of drug binding and recognition in relation to aldose reductase. L-Lyxose is also utilized by Escherichia coli, encoding a kinase for L-Xylulose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts as a reducing carbohydrate present in maple syrup. It is used in molecular modeling calculations in the study of drug binding and recognition in relation to aldose reductase. L-Lyxose is also utilized by Escherichia coli, encoding a kinase for L-Xylulose.
Solubility
Very soluble (586 g/L) (25°C) in water.
Notes
Hygroscopic. Store away from oxidizing agents, and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 108°C to 112°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Very soluble (586g/L) (25°C) in water.
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material L-Lyxose