Learn More
l-Lysine Monohydrochloride, Millipore™
Product Code. 10722262
Change view
Click to view available options
Quantity:
1 kg
10 kg
Unit Size:
10kg
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10722262 10 kg 10kg
10427690 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10722262 Supplier Millipore Supplier No. 1057009010
 Restricted Item
Estimated Shipment: 12-11-2026
For biochemistry
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 657-27-2
Specifications
Melting Point 262°C
MDL Number MFCD00064564
Synonym L-Lysine monohydrochloride, Lys, L-Lysine monohydrochloride
Density 1.28 g/cm3 at 20°C
Molecular Weight (g/mol) 182.65
Quantity 10 kg
Grade Research
Chemical Name or Material L-Lysine Monohydrochloride