Learn More
l-Leucine, Millipore™
Product Code. 10434171
Change view
Click to view available options
Quantity:
250 g
10 kg
Unit Size:
10kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10434171 10 kg 10kg
10177740 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10434171 Supplier Millipore Supplier No. 1053609010
 Restricted Item
Estimated Shipment: 12-11-2026
For biochemistry
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61-90-5
Specifications
Density 1.17 g/cm3 at 20°C
Quantity 10 kg
Grade Research
Chemical Name or Material L-Leucine