missing translation for 'onlineSavingsMsg'
Learn More

γ-L-Glutamyl-p-nitroanilide Hydrochloride, MP Biomedicals™

Product Code. 11478121 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11478121

Brand: MP Biomedicals 0215149501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Soluble in water (5mg/mL - clear, light yellow solution) or 0.1N HCl (1% w/v - clear, yellowish solution); soluble in methanol (100mg/mL with slight heat and constant stirring)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67953-08-6
Molecular Formula C11H14ClN3O5
Molecular Weight (g/mol) 303.70
MDL Number MFCD00039054
InChI Key WMRCHNNHOJNRSL-UHFFFAOYNA-N
Synonym gamma-l-glutamyl-p-nitroanilide, n-4-nitrophenyl-l-glutamine monohydrochloride, l-2-amino-4'-nitroglutaranilic acid hydrochloride, 2-amino-4-4-nitrophenyl carbamoyl butanoic acid hydrochloride, l-, a-glutamyl-p-nitroanilide, l-glutamic acid, a-p-nitroanilide hydrochloride, l-glutamic acid gamma-p-nitroanilide hydrochlori, l-glutamic acid gamma-p-nitroanilide hydrochloride, gamma-glutamyl transpeptidase substrate
IUPAC Name N-(3-carbamoyl-1-carboxypropyl)-4-nitroanilinium chloride
SMILES [Cl-].NC(=O)CCC([NH2+]C1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Color Beige
Quantity 1 g
Formula Weight 303.7
Physical Form Powder
Chemical Name or Material γ-L-Glutamyl-p-nitroanilide Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.