Learn More
L-Glutamic acid dibenzyl ester p-toluenesulfonate, 98%, Thermo Scientific™
Product Code. 15467746
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467746 5 g 5g
15457746 1 g 1g
15477746 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15467746 Supplier Thermo Scientific Alfa Aesar Supplier No. H62697.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2791-84-6
Molecular Formula C26H29NO7S
Molecular Weight (g/mol) 499.578
MDL Number MFCD00038980
InChI Key HVZUAIVKRYGQRM-LMOVPXPDSA-N
Synonym h-glu obzl-obzl.tosoh, l-glutamic acid dibenzyl ester tosylate, s-dibenzyl 2-aminopentanedioate 4-methylbenzenesulfonate, h-glu obzl-obzl p-tosylate, l-glutamic acid dibenzyl ester 4-toluenesulfonate, glu obzl-obzl.tosoh, dibenzyl l-glutamate p-toluenesulfonate, h-glu obzl-obzl 4-tosylate, l-glutamic acid dibenzyl ester p-toluenesulfonate, h-glu obzl-obzl inverted exclamation mark currencytosoh
PubChem CID 13233366
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N
Specifications
Quantity 5 g
Formula Weight 499.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Glutamic acid dibenzyl ester p-toluenesulfonate