Learn More
L-Glutamic acid dibenzyl ester 4-toluenesulfonate, TRC
Product Code. 30240957
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240957 500 mg 500mg
30240962 1 g 1g
30240780 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30240957 Supplier Toronto Research Chemicals Supplier No. TRCG597093500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2791-84-6
Molecular Formula C26H29NO7S
Molecular Weight (g/mol) 499.58
Synonym L-Glutamic acid, 1,5-bis(phenylmethyl) ester, 4-methylbenzenesulfonate (1:1) (ACI), L-Glutamic acid, bis(phenylmethyl) ester, 4-methylbenzenesulfonate (9CI), Glutamic acid, dibenzyl ester, p-toluenesulfonate (6CI, 7CI), Glutamic acid, dibenzyl ester, p-toluenesulfonate, L- (8CI), L-Glutamic acid dibenzyl ester p-toluenesulfonate, L-Glutamic acid dibenzyl ester tosylate, Dibenzyl L-glutamate p-toluenesulfonate, Dibenzyl L-glutamate tosylate, H-L-Glu(Obzl)-Obzl tosylate salt, L-glutamic acid dibenzyl ester 4-toluenesulfonate
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;4-methylbenzenesulfonic acid
SMILES Cc1ccc(cc1)S(=O)(=O)O.N[C@@H](CCC(=O)OCc2ccccc2)C(=O)OCc3ccccc3
Specifications
InChI Formula InChI=1S/C19H21NO4.C7H8O3S/c20-17(19(22)24-14-16-9-5-2-6-10-16)11-12-18(21)23-13-15-7-3-1-4-8-15;1-6-2-4-7(5-3-6)11(8,9)10/h1-10,17H,11-14,20H2;2-5H,1H3,(H,8,9,10)/t17-;/m0./s1
Quantity 500 mg
Formula Weight 499.1665
Shipping Condition Room Temperature
Chemical Name or Material L-Glutamic acid dibenzyl ester 4-toluenesulfonate