Learn More
L-Glutamic Acid alpha-tert-Butyl Ester, TRC
Product Code. 30006336
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006336 1 g 1g
30004990 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30006336 Supplier Toronto Research Chemicals Supplier No. TRCG5969801G
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 45120-30-7
Molecular Formula C9 H17 N O4
Molecular Weight (g/mol) 203.24
Synonym L-Glutamic Acid 1-(1,1-Dimethylethyl) Ester, L-Glutamic Acid tert-Butyl Ester, α-tert-Butyl L-Glutamate;
IUPAC Name (4S)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)[C@@H](N)CCC(=O)O
Specifications
InChI Formula InChI=1S/C9H17NO4/c1-9(2,3)14-8(13)6(10)4-5-7(11)12/h6H,4-5,10H2,1-3H3,(H,11,12)/t6-/m0/s1
Quantity 1 g
Formula Weight 203.1158
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Glutamic Acid alpha-tert-Butyl Ester
Grade HPLC