missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid, ≥99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
0.1kg
1kg
5kg
Description
Molecular biology, Cell Biology
Specifications
Specifications
| Color | White |
| Assay Percent Range | ≥99% |
| Density | 1.538g/mL at 20°C (Lit.) |
| IUPAC Name | 2-aminopentanedioic acid |
| Quantity | 1 kg |
| Formula Weight | 147.1 |
| Physical Form | Powder |
| Chemical Name or Material | L-Glutamic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction