missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, Cell Culture Reagent

Product Code. 15434209
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
500g
This item is not returnable. View return policy

Product Code. 15434209

Brand: Thermo Scientific Alfa Aesar J61651.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol

L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.

Solubility
It is soluble in water and HCL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
MDL Number MFCD00064228
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
PubChem CID 67678
ChEBI CHEBI:16283
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Melting Point >240°C (decomposition)
Color White
Beilstein 1728094
Merck Index 14,2782
Solubility Information It is soluble in water and HCL.
Density 1.68
Quantity 100 g
Formula Weight 240.3
Optical Rotation −219° (c=1 in 1N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Cystine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.