Learn More
l-Cysteine, Millipore™
Product Code. 10596632
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
10 kg
Unit Size:
100g
10kg
1kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10596632 1 kg 1kg
10307140 25 g 25g
10112920 100 g 100g
10433781 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10596632 Supplier Millipore Supplier No. 1028381000
 Restricted Item
Estimated Shipment: 12-11-2026
For biochemistry
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52-90-4
Specifications
Melting Point 220°C to 228°C
MDL Number MFCD00064306
Synonym L-Cysteine, α-Amino-β-mercapto propionic acid, αlpha-Amino-β-mercapto propionic acid, Cys, αlpha-Amino-beta-mercapto propionic acid, L-Cysteine
Molecular Weight (g/mol) 121.16
Quantity 1 kg
Grade Research
Chemical Name or Material L-Cysteine