missing translation for 'onlineSavingsMsg'
Learn More

L-Citrulline, 99.3%, MP Biomedicals™

Product Code. 11403350 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
1 kg
Unit Size:
1kg
25g
500g
5g
Product Code. Quantity unitSize
11403350 25 g 25g
11493340 5 g 5g
11423350 500 g 500g
11433350 1 kg 1kg
4 options
This item is not returnable. View return policy

Product Code. 11403350

Brand: MP Biomedicals 0210139725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
SMILES NC(CCCNC(N)=O)C(O)=O

Specifications

Color White
Assay Percent Range 99.3%
IUPAC Name 2-amino-5-(carbamoylamino)pentanoic acid
Quantity 25 g
Formula Weight 175.19
Physical Form Crystalline Powder
Chemical Name or Material L-Citrulline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.