Learn More
L-(-)-Bornyl acetate, 95%
Product Code. 11486106
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486106 25 g 25g
11496106 100 g 100g
11406116 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11486106 Supplier Thermo Scientific Alfa Aesar Supplier No. B22957.14
Request Bulk or Custom Format
CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5655-61-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135942,MFCD00135942
InChI Key KGEKLUUHTZCSIP-HOSYDEDBSA-N
Synonym bornyl acetate, -bornyl acetate
PubChem CID 44630108
SMILES CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
Specifications
Density 0.98
Boiling Point 223°C to 224°C
Flash Point 84°C (184°F)
Refractive Index 1.463
Quantity 25 g
Beilstein 2502036
Merck Index 14,1339
IUPAC Name (1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
Formula Weight 196.29
Percent Purity 95%
Chemical Name or Material L-(-)-Bornyl acetate