Learn More
L-Bornyl acetate, 95%
Product Code. 10254080 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10254080 5 g 5g
10060570 100 g 100g
10599800 500 g 500g
10217313 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10254080 Supplier Thermo Scientific Acros Supplier No. 106550050
Request Bulk or Custom Format
CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5655-61-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135942,MFCD00135942
InChI Key KGEKLUUHTZCSIP-HOSYDEDBSA-N
Synonym bornyl acetate, -bornyl acetate
PubChem CID 44630108
IUPAC Name [(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
SMILES CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
Specifications
Color Colorless
Density 0.9875g/mL
Boiling Point 223°C to 224°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (as C12H20O2) Total esters (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.465
Quantity 5 g
Beilstein 06,82
Merck Index 15,1341
Specific Gravity 0.9875
Specific Rotation -38
Solubility Information Solubility in water: .. Other solubilities: miscible with 95% alcohol and ether
Formula Weight 196.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material L-Bornyl acetate, 95%