Learn More
l-Alanine, Millipore™
Product Code. 10336560
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10336560 100 g 100g
10295150 25 g 25g
10188200 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10336560 Supplier Millipore Supplier No. 1010070100
 Restricted Item
Estimated Shipment: 12-11-2026
For biochemistry
for biochemistry
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56-41-7
Specifications
Melting Point 295°C to 297°C (decomposition)
MDL Number MFCD00166369
Synonym L-Alanine, 2-Aminopropionic acid, Ala, L-Alanine
Density 1.40 g/cm3 at 20°C
Molecular Weight (g/mol) 89.09
Quantity 100 g
Grade Research
Chemical Name or Material L-Alanine