Learn More
L-Alanine-d7, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | L-Alanine-d7 |
| CAS | 74280-71-0 |
| Molecular Formula | C3 D7 N O2 |
| Quantity | 0.5 g |
| Synonym | L-Alanine-N,N,1,2,3,3,3-d7 (ACI), L-Alanine-d7 (9 CI), Alanine-d7 (6 CI), (2 S)-2-Aminopropanoic-d7 acid, L-Alanine-N,N,O,2,3,3,3-d7, L-Alanine-D7 |
| InChI Formula | InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i1D3,2 D/hD3 |
| SMILES | [2 H]OC(=O)[C@@]([2 H])(N([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | deuterio (2 S)-2,3,3,3-tetradeuterio-2-(dideuterioamino)propanoate |
| Molecular Weight (g/mol) | 96.1363 |
| Formula Weight | 96.0916 g/mol |
| Show More |