Learn More
L-Alanine 2-ethylbutyl ester hydrochloride, Thermo Scientific™
Product Code. 17740709 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
17740709 5 g 5g
17730709 1 g 1g
2 options
Product Code. 17740709
Brand: Thermo Scientific Acros 469010050
Request Bulk or Custom Format
Specifications
Alpha Vector ALANINEETHYLBUTYLESTERHYDROCHLORIDE
Chemical Name or Material L-Alanine 2-ethylbutyl ester hydrochloride
CAS 946511-97-3
Melting Point 75°C to 80°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C9 H19 N O2 . H Cl
MDL Number 32660031
Quantity 5 g
Synonym (S)-2-Ethylbutyl 2-aminopropanoate hydrochloride
Molecular Weight (g/mol) 209.74
Physical Form Crystalline Powder