missing translation for 'onlineSavingsMsg'
Learn More

L-779450, MedChemExpress

Product Code. p-200096450
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16491537 50 mg 50mg
16475976 100 mg 100mg
16475348 10 mg 10mg
16448067 5 mg 5mg
4 options
This item is not returnable. View return policy

Product Code. 16491537

Brand: Medchem Express HY12787

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-779450 is a potent and selective B-Raf kinase inhibitor with a Kd of 2.4 nM.

L-779450 is a potent and selective B-Raf kinase inhibitor with a Kd of 2.4 nM.

Specifications

Chemical Name or Material L-779450
CAS 303727-31-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H14ClN3O
Quantity 50 mg
Solubility Information DMSO : ≥ 34 mg/mL (97.76 mM)
SMILES OC1=CC(C2=C(C3=CC=NC=C3)N=C(C4=CC=CC=C4)N2)=CC=C1Cl
Molecular Weight (g/mol) 347.8
Formula Weight 347.8
Percent Purity 98.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.