missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-2-Phenylglycine methyl ester hydrochloride, 97%

Product Code. p-7041628
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15437946

Brand: Thermo Scientific Alfa Aesar H62874.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15028-39-4 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15028-39-4
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00077158
InChI Key DTHMTBUWTGVEFG-QRPNPIFTSA-N
Synonym h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
PubChem CID 11217910
IUPAC Name methyl (2S)-2-amino-2-phenylacetate;hydrochloride
SMILES COC(=O)C(C1=CC=CC=C1)N.Cl

Specifications

Melting Point 200°C (decomposition)
Quantity 25 g
Formula Weight 201.65
Optical Rotation +118° (c=1 in Water)
Percent Purity 97%
Chemical Name or Material L-(+)-2-Phenylglycine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.