Learn More
Saint Gobain Life Sciences Kynar Bag
Product Code. 13305211
Change view
Click to view available options
Capacity (Metric):
7 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
13305211 7 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13305211 Supplier Saint Gobain Life Sciences Supplier No. 0141015
  • ONLINE EXCLUSIVE PRICE
    1045.00 NOK
    1017.29 NOK / Pack of 1
    Save 27.71 NOK
    2% Off
Estimated Shipment: 20-01-2027
Use this Kynar Bag for gas sample analysis. Rubber gas sample bag features corrosive-resistant Kynar construction for all your application needs.
  • Strong seams for added durability
  • Single-wide seam styles
  • Available in 2 mil and 4 mil thick
  • Individually sealed in a dust-protected outer bag that also blocks out sunlight
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Gas Sample Analysis
Material Rubber
Thickness (English) 0.003 in.
Dimensions (L x W) 610 x 610 mm
Length (English) 24 in.
Width (English) 24 in.
Product Type Kynar Bag
Capacity (English) 1.8 gal.
Capacity (Metric) 7 L
Length (Metric) 610 mm
Width (Metric) 610 mm
Thickness (Metric) 0.076 mm