Learn More
KW-2478, MedChemExpress
Product Code. 16458397
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16458397
Brand: Medchem Express HY13468
MedChemExpress KW-2478 is an inhibitor of Hsp90α, with an IC50 of 3.8 nM, and has antitumor activity against various human hematological tumor cells.
KW-2478 is an inhibitor of Hsp90α, with an IC50 of 3.8 nM, and has antitumor activity against various human hematological tumor cells.
Specifications
Chemical Name or Material KW-2478
CAS 819812-04-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H42N2O9
Quantity 5 mg
Solubility Information DMSO : 200 mg/mL (348.03 mM; Need ultrasonic)
SMILES O=C(N(CCOC)CCOC)CC1=C(C(C2=CC=C(OCCN3CCOCC3)C(OC)=C2)=O)C(O)=CC(O)=C1CC
Molecular Weight (g/mol) 574.66
Formula Weight 574.66
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.