Learn More
KTN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19841-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody reacts with normal gastrointestinal epithelium as well as gastrointestinal and breast tumors, resulting in a cytoplasmic staining pattern.
This antibody reacts with normal gastrointestinal epithelium as well as gastrointestinal and breast tumors, resulting in a cytoplasmic staining pattern.Specifications
| KTN1 | |
| Polyclonal | |
| Unconjugated | |
| KTN1 | |
| CG 1 antigen, CG1, KIAA0004, Kinectin, kinectin 1 (kinesin receptor), Kinesin receptor, KNT, KTN1, MU RMS 40.19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16709, 3895 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q61595, Q86UP2 | |
| KTN1 | |
| KTN1 Fusion Protein Ag13854 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |