Learn More
Klever KS Series Cutter
Product Code. 17739017
Change view
Click to view available options
Blade Material:
Stainless Steel
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Blade Material Unit Size
17739017 Stainless Steel Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17739017 Supplier Klever Supplier No. KS101SSW
  • ONLINE EXCLUSIVE PRICE
    2098.05 NOK
    1619.08 NOK / Pack of 5
    Save 478.97 NOK
    22% Off
View all promotions
Estimated Shipment: 19-01-2027
Klever KS Series Cutter with Wide Opening Stainless Steel Handle and Carriage.
  • Do you want a safe cutting tool that is particularly easy to disassemble because it consists of only two parts (plus blade)? Are you looking for a safety knife that is particularly easy to clean? Then the Klever KS101SS-W safety knife is the one for you.
    • It cuts up to 1.5-wall corrugated cardboard, but also almost any other material.
    • Suitable for auto-clave cleaning
Specifications
Blade Material Stainless Steel
Product Type Cutter
Disposable Reusable
Sterility Non-sterile
Color Silver