Learn More
IKA™ KS 3000 ic Control
Innovative incubator shaker design to allow for unattended operation in a temperature-controlled environment.
83655.00 NOK
Specifications
Speed Range 10 to 500 rpm
Voltage 100/115/230 V
Temperature Range (Metric) Ambient to 80°C
Wattage 1120 W
Frequency 50/60 Hz
View More Specs
Products 1
Product Code Brand Plug Type Price Quantity & Availability  
Product Code Brand Plug Type Price Quantity & Availability  
16288161
View Documents Promotion Details
IKA™
0003940100
EU Plug
83655.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-08-2024
Description
  • Large LED display for speed and time settings
  • Controls with antimicrobial coating for reduction of bacteria
  • Integrated PID temperature control (use of two PT 1000 temperature sensors)
  • Junction box in the workspace for connection of an additional temperature sensor e.g. PT 1000.60
  • Electronic temperature and speed control
  • Electronic timer switch: 0 to 999 hr (set by the minute or by the hour)
  • Optional built-in cooler for connection to an external cooling unit e.g. RC 2 basic / control
  • Unit switches off automatically if disturbed
  • Unit stops automatically when hood is lifted
  • Collecting tray with drain hose on rear of unit
  • Simple operation
  • All functions can be controlled and documented with labworldsoft™
  • Shaking movement: orbital
  • Orbital diameter: 20 mm
Specifications
Specifications
10 to 500 rpm
Ambient to 80°C
50/60 Hz
37 kg
80%
100/115/230 V
1120 W
258 x 330 x 330 mm
LED
1 min. to 999 hr.