Learn More
IKA™ KS 260 Control NOL Flat Shaker
Product Code. 15895931 Shop All IKA Products
Change view
Click to view available options
Speed Range:
0 to 500 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
15895931 0 to 500 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15895931 Supplier IKA™ Supplier No. 0004278400
  • ONLINE EXCLUSIVE PRICE
    42385.00 NOK
    41121.21 NOK / Pack of 1
    Save 1263.79 NOK
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Compact, flat shaker with ideal swivel motion.
  • Electronic adjustment of speed and timer
  • Digital display makes it possible to read the speed, timer function and operating modes
  • Electronic time switching clock
  • All functions can be controlled and documented with labworldsoft™ software
  • Special version with reverse rotating direction on request
  • Wide range of attachment combinations makes it possible to use almost all shapes and sizes of vessels
  • Attachments are not included, please order separately
TRUSTED_SUSTAINABILITY
Specifications
Orbit 10 mm
Load Bearing Capacity (Metric) 7.5 kg
Speed Range 0 to 500 rpm
Electrical Requirements 220/240 V, 100/120 V, 50/60 Hz, 45 W
Wattage 45 W
Frequency 50/60 Hz
Dimensions (L x W x H) 360 x 98 x 420 mm
Weight (Metric) 8.8 kg
Display LCD: Speed, Timer
Includes RS 232 interface
Timer Range 1 to 599 min.