Learn More
KS 100μS Test and Calibration Standards, WTW™
Click to view available options
Quantity:
250 mL
Unit Size:
300mL
Description
- Test and calibration standard 100 μS/cm at 25 °C
- Test resistors and calibration solutions to test the meters with conductivity function as well as to calibrate the system from the device and conductivity measuring cell
- Optimal measuring safety by PTB/NIST traceable standard solutions
- Easy device check by highly precise test resistors
- Pharmacopeia conform test kits
- The system including measuring
Specifications
Specifications
| Packaging | Borosilicate Glass |
| Density | 1.2g/cm3 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 120°C |
| Quantity | 250 mL |